Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alpha-thujone | 546-80-5 | 98.1% | 100 MG
SDP

Catalog No. 5000230938
Encompass_Preferred

Alpha-thujone (CAS 546-80-5) is a monoterpene ketone research chemical that modulates GABAA receptor activity. The compound has formula C10H16O, molecular weight 152.24, and is supplied as a high-purity reference standard for laboratory research.

  • Monoterpene ketone with GABAA receptor activity.
  • Molecular weight 152.24 and formula C10H16O.
  • Typical purity reported as 98.07% for the product in supplier documentation.
  • Recommended storage: pure form -20°C for long-term, 4°C for shorter-term; in solvent keep at -80°C or -20°C depending on duration.
  • For research use only; not for human consumption.

Catalog No. 50-002-30938 Supplier Medchemexpress LLC Supplier No. HY121618100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alpha-thujone (CAS 546-80-5) is a monoterpene ketone research chemical that modulates GABAA receptor activity. The compound has formula C10H16O, molecular weight 152.24, and is supplied as a high-purity reference standard for laboratory research.

  • Monoterpene ketone with GABAA receptor activity.
  • Molecular weight 152.24 and formula C10H16O.
  • Typical purity reported as 98.07% for the product in supplier documentation.
  • Recommended storage: pure form -20°C for long-term, 4°C for shorter-term; in solvent keep at -80°C or -20°C depending on duration.
  • For research use only; not for human consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.