Learn More
Sigma Aldrich Fine Chemicals Biosciences alpha Tocopherol phenyl 131MG
Catalog No. 502908414
Encompass
Tocopherol (vitamin E) is a lipid-soluble antioxidant found in whole grains nuts andnbspoils. It is known to scavenge reactive oxygen species by interacting with lipids duringnbspoxidative stress.Tocopherol-(phenyl-13C6) is an isotope of vitamin E wherein all the six phenyl carbons are replaced by 13C6 carbon.
Catalog No. 50-290-8414 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 9003741MG
Tocopherol (vitamin E) is a lipid-soluble antioxidant found in whole grains nuts andnbspoils. It is known to scavenge reactive oxygen species by interacting with lipids duringnbspoxidative stress.Tocopherol-(phenyl-13C6) is an isotope of vitamin E wherein all the six phenyl carbons are replaced by 13C6 carbon.