Learn More
TARGETMOL CHEMICALS INC ALPHA-TOCOPHEROL PHOSPHA 5MG
SDP
Catalog No. 502579393
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. (alpha)-Tocopherol phosphate (alpha-Tocopherol phosphate) is an antioxidant that scavenges UVA1-induced reactive oxygen species (ROS) and protects against long-wave UVA1-induced cell death. alpha-Tocopherol phosphate disodium impedes endothelial progenitor cell apoptosis enhances endothelial progenitor cell migration under high glucose/low oxygen conditions and promotes angiogenesis. (alpha)-Tocopherol phosphate disodium (Vitamin E phosphate disodium) prevents apoptosis enhances migration of endothelial progenitor cells under high glucose/low oxygen conditions and promotes angiogenesis. purity: 99%
Catalog No. 50-257-9393 Supplier TARGETMOL CHEMICALS INC Supplier No. T639185MG
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. (alpha)-Tocopherol phosphate (alpha-Tocopherol phosphate) is an antioxidant that scavenges UVA1-induced reactive oxygen species (ROS) and protects against long-wave UVA1-induced cell death. alpha-Tocopherol phosphate disodium impedes endothelial progenitor cell apoptosis enhances endothelial progenitor cell migration under high glucose/low oxygen conditions and promotes angiogenesis. (alpha)-Tocopherol phosphate disodium (Vitamin E phosphate disodium) prevents apoptosis enhances migration of endothelial progenitor cells under high glucose/low oxygen conditions and promotes angiogenesis. purity: 99%