Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alpha-tocopherol phosphate | 38976-17-9 | >98.0% | 510.69 | C29H51O5P | 10 MG
SDP

Catalog No. 5000289244
Encompass_Preferred

Alpha-tocopherol phosphate is a phosphorylated derivative of vitamin E supplied as a high-purity analytical standard for biochemical research. It is provided as a small-scale powder with documented solubility and storage recommendations for laboratory use.

  • High-purity analytical standard suitable for biochemical assays (purity >98%).
  • Functions as an antioxidant and vitamin E derivative.
  • Supplied as a 10 mg powder for small-scale experiments.
  • Molecular weight 510.69 and formula C29H51O5P.
  • Soluble in DMSO (≥100 mg/mL) and has low aqueous solubility.
  • Stable when stored at -20°C as a powder with extended shelf life.
  • Intended for research use only; not for clinical or human use.

Catalog No. 50-002-89244 Supplier Medchemexpress LLC Supplier No. HY1668610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alpha-tocopherol phosphate is a phosphorylated derivative of vitamin E supplied as a high-purity analytical standard for biochemical research. It is provided as a small-scale powder with documented solubility and storage recommendations for laboratory use.

  • High-purity analytical standard suitable for biochemical assays (purity >98%).
  • Functions as an antioxidant and vitamin E derivative.
  • Supplied as a 10 mg powder for small-scale experiments.
  • Molecular weight 510.69 and formula C29H51O5P.
  • Soluble in DMSO (≥100 mg/mL) and has low aqueous solubility.
  • Stable when stored at -20°C as a powder with extended shelf life.
  • Intended for research use only; not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.