Learn More
U.S. Pharmacopeia Alpha Tocopheryl Acetate, 7695-91-2, MFCD00072042, 250mg
Catalog No. NC9199955
Encompass
Molecular Formula C31H52O3, Molecular Weight 472.749, Melting Point 79.7 - 81.5 °F (26.5 - 27.5 °C), Synonyms: Vitamin E acetate, 2H-1-Benzopyran-6-ol, 3,4-dihydro-2,5,7,8-tetramethyl-2-[4,8,12-trimethyltidecyl)-, acetate; Cold Shipment Required.
Catalog No. NC9199955 Supplier U.S. Pharmacopeia Supplier No. 1667701
Molecular Formula C31H52O3, Molecular Weight 472.749, Melting Point 79.7 - 81.5 °F (26.5 - 27.5 °C), Synonyms: Vitamin E acetate, 2H-1-Benzopyran-6-ol, 3,4-dihydro-2,5,7,8-tetramethyl-2-[4,8,12-trimethyltidecyl)-, acetate; Cold Shipment Required.