Learn More
Abcam alpha-Tocotrienol, Form of vitamin E, 25MG
Catalog No. 502705702
Encompass_Preferred
MW 424.7 Da, Purity >98%. Form of vitamin E. Lipophilic antioxidant agent. Tocotrienol β (ab145176), δ (ab144449) and γ (ab144448) analog. Induces apoptosis in cancer cells. Shows antitumor, neuroprotective and antioxidant effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5702 Supplier Abcam Supplier No. AB14445125MG
MW 424.7 Da, Purity >98%. Form of vitamin E. Lipophilic antioxidant agent. Tocotrienol β (ab145176), δ (ab144449) and γ (ab144448) analog. Induces apoptosis in cancer cells. Shows antitumor, neuroprotective and antioxidant effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement