Learn More
Med Vet International PHV, ZOETIS, ALPHA TRAK 2 STARTER KIT
Catalog No. 502188510
Encompass_Preferred
- Improved test strip design for quick and easy wicking (angled sample area makes it easy to position the target area to the blood drop) - Meter turns on and off automatically as strip is inserted/removed - New AlphaTRAKer Electronic Data Master allows pet owners to record information for their pets, providing you with easy to read charts and graphs of test results. - New lower every day starter kit price. * Starter kit includes: AlphaTRAK 2 Meter, 25 Test Strips, Lancing Device, Instructional DVD, Diabetes Diary, Carrying Case, User’s Guide, 30 Lancets, Control Solution.
Catalog No. 50-218-8510 Supplier Med Vet International Supplier No. PHALPHATRAKKIT
- Improved test strip design for quick and easy wicking (angled sample area makes it easy to position the target area to the blood drop) - Meter turns on and off automatically as strip is inserted/removed - New AlphaTRAKer Electronic Data Master allows pet owners to record information for their pets, providing you with easy to read charts and graphs of test results. - New lower every day starter kit price. * Starter kit includes: AlphaTRAK 2 Meter, 25 Test Strips, Lancing Device, Instructional DVD, Diabetes Diary, Carrying Case, User’s Guide, 30 Lancets, Control Solution.