Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alpha Tubulin (acetyl K40) Antibody (YA626) | 55 kDa (observed band size) | 10 UL
SDP

Catalog No. 5000396222
Encompass_Preferred

Alpha Tubulin (acetyl K40) Antibody (YA626) is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting alpha Tubulin (acetyl K40). This antibody is protein A affinity purified and is suitable for research applications, including Western Blot (WB).

  • Host: Rabbit
  • Clonality: Monoclonal
  • Target: Alpha Tubulin (acetyl K40)
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Application: WB (1:1000 dilution)
  • Storage: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Formulation: Supplied in 1xPBS (pH7.4), 0.1% BSA, 40% Glycerol, with 0.05% Sodium Azide as preservative

Catalog No. 50-003-96222 Supplier Medchemexpress LLC Supplier No. HYP8048510UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alpha Tubulin (acetyl K40) Antibody (YA626) is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting alpha Tubulin (acetyl K40). This antibody is protein A affinity purified and is suitable for research applications, including Western Blot (WB).

  • Host: Rabbit
  • Clonality: Monoclonal
  • Target: Alpha Tubulin (acetyl K40)
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Application: WB (1:1000 dilution)
  • Storage: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Formulation: Supplied in 1xPBS (pH7.4), 0.1% BSA, 40% Glycerol, with 0.05% Sodium Azide as preservative

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.