Learn More
LICOR Tubulin Mouse Monoclonal Antibody for Normalization (100 L)
Supplier:  LICOR 92642213
Encompass_Preferred
α-Tubulin primary antibody can be used as an internal loading control (ILC) for normalization. The expression of α-tubulin, or any housekeeping protein (HKP), should be validated to ensure that its expression does not change under experimental conditions. Once validated, α-tubulin primary antibodies can be used for the detection of α-tubulin when performing two-color detection. Detect α-Tubulin Mouse Monoclonal Antibody with IRDye Goat anti-Mouse or IRDye Donkey anti-Mouse secondary antibodies.
Catalog No. NC0244815