Learn More
Description
- α-vinylbenzyl alcohol is an allyl alcohol derivative
- Its isomerization reaction using a novel ternary catalyst system has been reported
- The kinetic resolution of racemic α-vinylbenzyl alcohol by tungsten-catalyzed asymmetric epoxidation method has been repoorted
- The coupling reaction of α-vinylbenzyl alcohol and aldehydes in the presence of rucl2(pph3)3 catalyst and in(oac)3 as a co-catalyst in ionic liquids has been investigated
- Etherification reaction of glycerol with α-vinylbenzyl alcohol in the presence of sulfonic catalyst immobilized on silica (sio2-so3h) has been reported.αvinylbenzyl alcohol may be used in the synthesis of cinnamyl ethers by reacting with alcohols
Specifications
Specifications
| CAS | 42273-76-7 |
| Density | 1.021 g/mL (at 25°C (literature)) |
| Boiling Point | 101°C to 102°C (12 mmHg) |
| Molecular Formula | C9H10O |
| Refractive Index | n20/D 1.54 (literature) |
| Linear Formula | C6H5CH(CH=CH2)OH |
| MDL Number | MFCD00093987 |
| Quantity | 10 g |
| Molecular Weight (g/mol) | 134.18 |
| Percent Purity | 97% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.