Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich alpha-Vinylbenzyl alcohol

Catalog No. 4237691G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-376-91G 1 g
42-376-910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-376-91G Supplier Sigma Aldrich Supplier No. 4237691G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • α-vinylbenzyl alcohol is an allyl alcohol derivative
  • Its isomerization reaction using a novel ternary catalyst system has been reported
  • The kinetic resolution of racemic α-vinylbenzyl alcohol by tungsten-catalyzed asymmetric epoxidation method has been repoorted
  • The coupling reaction of α-vinylbenzyl alcohol and aldehydes in the presence of rucl2(pph3)3 catalyst and in(oac)3 as a co-catalyst in ionic liquids has been investigated
  • Etherification reaction of glycerol with α-vinylbenzyl alcohol in the presence of sulfonic catalyst immobilized on silica (sio2-so3h) has been reported.αvinylbenzyl alcohol may be used in the synthesis of cinnamyl ethers by reacting with alcohols

Specifications

CAS 42273-76-7
Density 1.021 g/mL (at 25°C (literature))
Boiling Point 101°C to 102°C (12 mmHg)
Molecular Formula C9H10O
Refractive Index n20/D 1.54 (literature)
Linear Formula C6H5CH(CH=CH2)OH
MDL Number MFCD00093987
Quantity 1 g
Molecular Weight (g/mol) 134.18
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.