Learn More
New England Biolabs, Inc. α2-3,6,8,9 Neuraminidase A - 800 units
SDP
Catalog No. NC0762323
Encompass
a2-3,6,8,9 Neuraminidase A is a broad specificity sialidase, which cleaves linear and branched non-reducing terminal sialic acid residues from glycoproteins, glycopeptides, and oligosaccharides. It can be used for glycan analysis and characterization and intact glycoproteins.
Catalog No. NC0762323 Supplier New England Biolabs, Inc. Supplier No. P0722S
a2-3,6,8,9 Neuraminidase A is a broad specificity sialidase, which cleaves linear and branched non-reducing terminal sialic acid residues from glycoproteins, glycopeptides, and oligosaccharides. It can be used for glycan analysis and characterization and intact glycoproteins.