Learn More
Sigma Aldrich Fine Chemicals Biosciences alpha2 hs Glycoprotein fro1MG
Catalog No. 502904925
Encompass
Î2-HS glycoprotein (A2HS) is a glycoprotein (molecular weight 49 000) present in human plasma at levels ranging between 40 and 85 mg/dl. A2HS is a major protein component of the bone matrix and could be involved both in mineralization and bone resorption.
Catalog No. 50-290-4925 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G05161MG
Î2-HS glycoprotein (A2HS) is a glycoprotein (molecular weight 49 000) present in human plasma at levels ranging between 40 and 85 mg/dl. A2HS is a major protein component of the bone matrix and could be involved both in mineralization and bone resorption.