Learn More
Enzo Life Sciences alphaA/alphaB Crystallin polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA224F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human αA- and αB-Crystallin. Source: Purified from rabbit serum. Species reactivity: Human, Mouse Bovine. Applications: ICC, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide and 50% glycerol.
Catalog No. 50-200-7420