Learn More
Neuromics, Inc Alphabiocoat Solution, 20 mL
Catalog No. NC1045480
Encompass
AlphaBioCoat Solution (AC001) is a biocompatible complex of extracellular matrix binding solution that is supplemented with growth factors. It helps accelerate cell attachment and cell growth.
Catalog No. NC1045480 Supplier Neuromics, Inc Supplier No. AC001
AlphaBioCoat Solution (AC001) is a biocompatible complex of extracellular matrix binding solution that is supplemented with growth factors. It helps accelerate cell attachment and cell growth.