Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alphadolone acetate | 23930-37-2 | 98.5% | 390.51 | C23H34O5 | 10 MG
SDP

Catalog No. 5000243043
Encompass_Preferred

Alphadolone acetate is a steroid anesthetic research compound (CAS 23930-37-2) used in pharmacology and nociception studies. It is provided as a purified small-molecule reagent intended for in vitro and in vivo experimental applications.

  • Steroid anesthetic with analgesic and antinociceptive activity.
  • High purity: 98.5%.
  • Molecular formula C23H34O5; molecular weight 390.51 g/mol.
  • Supplied in multiple small-mass pack sizes suitable for research dosing.
  • Soluble in DMSO for biological assay preparation.

Catalog No. 50-002-43043 Supplier Medchemexpress LLC Supplier No. HY10806810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alphadolone acetate is a steroid anesthetic research compound (CAS 23930-37-2) used in pharmacology and nociception studies. It is provided as a purified small-molecule reagent intended for in vitro and in vivo experimental applications.

  • Steroid anesthetic with analgesic and antinociceptive activity.
  • High purity: 98.5%.
  • Molecular formula C23H34O5; molecular weight 390.51 g/mol.
  • Supplied in multiple small-mass pack sizes suitable for research dosing.
  • Soluble in DMSO for biological assay preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.