Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alphadolone acetate | 23930-37-2 | MFCD00200116 | 98.5% | 390.51 | C23H34O5 | 25 MG
SDP

Catalog No. 5000243032
Encompass_Preferred

Alphadolone acetate is a steroid anesthetic research chemical with reported antinociceptive and analgesic properties. It is supplied as a solid for laboratory preparation and handling, intended for research use only.

  • Steroid anesthetic with antinociceptive activity.
  • High purity (98.5%).
  • Molecular formula C23H34O5 and molecular weight 390.51.
  • Soluble in DMSO; recommended formulation concentration 250 mg/mL.
  • Supplied in milligram quantities for laboratory use.
  • Storage: powder at -20°C for long term or 4°C for short term; solutions at -80°C for long term.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-43032 Supplier Medchemexpress LLC Supplier No. HY10806825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alphadolone acetate is a steroid anesthetic research chemical with reported antinociceptive and analgesic properties. It is supplied as a solid for laboratory preparation and handling, intended for research use only.

  • Steroid anesthetic with antinociceptive activity.
  • High purity (98.5%).
  • Molecular formula C23H34O5 and molecular weight 390.51.
  • Soluble in DMSO; recommended formulation concentration 250 mg/mL.
  • Supplied in milligram quantities for laboratory use.
  • Storage: powder at -20°C for long term or 4°C for short term; solutions at -80°C for long term.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.