Learn More
Sino Biological alpha-Galactosidase A / GLA Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 12078R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human alpha-Galactosidase A / GLA (rh alpha-Galactosidase A / GLA; Catalog#12078-H08H; NP_000160.1; Met 1-Leu 429).
Catalog No. 50-224-7424