Learn More
Revvity Health Sciences Inc Triiiodothyronine, L-3, 5, 3'-[125I], 500 µCi (18.5 MBq), Specific Activity: 3300 µCi (122 MBq)/µg
Catalog No. 502812391
Encompass
Triiiodothyronine, L-3, 5, 3'-[125I], 500 µCi (18.5 MBq), Specific Activity: 3300 µCi (122 MBq)/µg
Catalog No. 50-281-2391 Supplier Revvity Health Sciences Inc Supplier No. NEX110X500UC
Triiiodothyronine, L-3, 5, 3'-[125I], 500 µCi (18.5 MBq), Specific Activity: 3300 µCi (122 MBq)/µg