Learn More
Revvity Health Sciences Inc Sulfur-35 Radionuclide, 25mCi (925MBq), Specific Activity: 1050-1600Ci (38.8-59.2TBq)/mmol, Sulfuric Acid in 1mL Water
Catalog No. 502812392
Encompass
Sulfur-35 Radionuclide, 25mCi (925MBq), Specific Activity: 1050-1600Ci (38.8-59.2TBq)/mmol, Sulfuric Acid in 1mL Water
Catalog No. 50-281-2392 Supplier Revvity Health Sciences Inc Supplier No. NEX042025MC
Add to Cart
Add to Cart
Request a Quote
Sulfur-35 Radionuclide, 25mCi (925MBq), Specific Activity: 1050-1600Ci (38.8-59.2TBq)/mmol, Sulfuric Acid in 1mL Water