Learn More
Revvity Health Sciences Inc Iodine-125 Radionuclide, 10mCi, 10-5M NaOH (pH 8-11), Concentration: >350mCi/mL
Supplier:  Revvity Health Sciences Inc NEZ033L010MC
Encompass
Iodine-125 Radionuclide, 10mCi, 10-5M NaOH (pH 8-11), Concentration: >350mCi/mL
Catalog No. 50-209-9745