Learn More
Revvity Health Sciences Inc L-[35S]-Methionine, 5mCi (185MBq), Specific Activity:>800Ci (29.6TBq)/mmole, 50mM Tricine, 10mM BME
Catalog No. 502812300
Encompass
L-[35S]-Methionine, 5mCi (185MBq), Specific Activity:>800Ci (29.6TBq)/mmole, 50mM Tricine, 10mM BME
Catalog No. 50-281-2300 Supplier Revvity Health Sciences Inc Supplier No. NEG009H005MC
L-[35S]-Methionine, 5mCi (185MBq), Specific Activity:>800Ci (29.6TBq)/mmole, 50mM Tricine, 10mM BME