Learn More
Revvity Health Sciences Inc Triiiodothyronine, L-3, 5, 3'-[125I], 100 µCi (3.7 MBq), Specific Activity: 3300 µCi (122 MBq)/µg
Catalog No. 502812217
Encompass
Triiiodothyronine, L-3, 5, 3'-[125I], 100 µCi (3.7 MBq), Specific Activity: 3300 µCi (122 MBq)/µg
Catalog No. 50-281-2217 Supplier Revvity Health Sciences Inc Supplier No. NEX110X100UC
Add to Cart
Add to Cart
Triiiodothyronine, L-3, 5, 3'-[125I], 100 µCi (3.7 MBq), Specific Activity: 3300 µCi (122 MBq)/µg