Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alphamine Red R Base 90.0+%, TCI America™
SDP

Catalog No. A04741G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A04741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A04741G Supplier TCI America Supplier No. A04741G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 57322-42-6
Molecular Formula C26H19N3O6S2
Molecular Weight (g/mol) 533.57
MDL Number MFCD00135326
InChI Key UMZJOYSJYIDKDN-UHFFFAOYSA-N
Synonym 3-(4-Anilino-1-naphthylazo)-2,7-naphthalenedisulfonic Acid
PubChem CID 407428
IUPAC Name 3-{2-[4-(phenylamino)naphthalen-1-yl]diazen-1-yl}naphthalene-2,6-disulfonic acid
SMILES OS(=O)(=O)C1=CC=C2C=C(C(=CC2=C1)N=NC1=C2C=CC=CC2=C(NC2=CC=CC=C2)C=C1)S(O)(=O)=O

Specifications

Quantity 1 g
Formula Weight 533.57
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Alphamine Red R Base
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.