Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaProTech Cleanroom MVT Face Masks
BIOPHARMA

Catalog No. 19125440 Shop All AlphaProTech Products
Change view
Click to view available options
Size:
Standard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-125-440 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-125-440 Supplier AlphaProTech Supplier No. 9060
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Face masks with earloops are designed to shield mouth while allowing ease of breathing.

  • Clean mask secured with soft knit earloops
  • Disposable
  • Soft
  • White

Specifications

Product Type Cleanroom Face Mask
Color White
Size Standard
Sterility Non-sterile
Style Pleated
Disposable Single-use
For Use With (Application) Semiconductor, Disk Drive, Medical Device, Pharmaceutical, Biomedical
Model 9060
Packaging Type Double-bagged, Cleanroom Packaged
Series MVT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.