Learn More
AlphaProTech Critical Cover™ AlphaAir™ Face Masks
BIOPHARMA
Catalog No. p-3548835 Shop All AlphaProTech Products
Change view
Click to view available options
Color:
Blue
Yellow
Fastening Type:
Earloops
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Fastening Type
19-130-4181 Blue Earloops
19-065-338 Yellow Earloops
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-130-4181 Supplier AlphaProTech Supplier No. BL 5005
Add to Cart
Add to Cart
This item is not returnable. View return policy
Constructed of cool, comfortable hypoallergenic materials that are naturally fluid-resistant. AlphaAir™ face masks feature a 0.1 micron filter with greater than 95% bacterial filtration efficiency.
  • Simple, cool and comfortable design
  • Constructed of materials that are naturally fluid-resistant
  • Highly malleable aluminum nosepiece extends entire length of mask, maximizing comfort and minimizing fogging of eyewear
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 3
Color Blue
Fastening Type Earloops
Material Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Construction 3-ply
Disposable Single-use
Filter Efficiency ≥98% (PFE), ≥98% (BFE)
Latex Free Yes
Model BL 5005
Pore Size 0.1 μm
Resistance Fluid
Series AlphaAir™
Quantity 500/Cs.