Learn More
AlphaProTech Critical Cover™ AlphaGuard™ Ear Loop Mask
Catalog No. 191304180 Shop All AlphaProTech Products
Change view
Click to view available options
Fastening Type:
Earloops
Catalog No. Fastening Type
19-130-4180 Earloops
1 options
This item is not returnable. View return policy
Catalog No. 19-130-4180 Supplier AlphaProTech Supplier No. 9050
  • $151.00
    $122.65 / Case of 500
    Save $28.35
    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
For superior comfort and protection
  • High breathability, fluid resistant
  • Excellent water vapor transmission rate and particle filtration to 0.1 micron.
  • Designed for use in cleanroom environments
  • Soft, latex-free, non-irritating knit ear loops are ultrasonically welded to the mask
Specifications
Product Type Cleanroom Face Mask
Color Blue
Fastening Type Earloops
Material Composite Fabric
Certifications/Compliance ASTM F2299
Cleanroom Class Class 100 (ISO 5)
Filter Efficiency 93.5% (PFE)
Latex Free Yes
Model 9050
Pore Size 0.1 μm (PFE)
Resistance Fluid