Learn More
AlphaProTech Critical Cover™ BarrierTech™ Gown
BIOPHARMA
Catalog No. 191694038 Shop All AlphaProTech Products
This item is not returnable. View return policy
Catalog No. 19-169-4038 Supplier AlphaProTech Supplier No. GN225243
Add to Cart
Add to Cart
This item is not returnable. View return policy
Provides protection for the wearer from particles, fluids and light chemical splash.
  • Durable and fluid resistant solution for lab and/or production areas
  • Low particle count for use in controlled environments
  • Banded ties attached to the neck and waist for easy and secure closure
  • Serged seams and elastic wrists
Specifications
No. of Pockets None
Style Full Coverage
Length (English) 45 in.
Gender Unisex
Packaging Type Bulk
For Use With (Application) Controlled Environment
Material Polyethylene, Polypropylene
Closure Type Neck and Waist Ties
Disposable Single-use
Sterility Non-sterile
Latex Free Yes
Seam Type Serged
Sleeve Style Long Sleeve
Certifications/Compliance 21 CFR Part 820
Autoclavable Not Autoclavable