Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaProTech Critical Cover™ ComforTech™ Sleeves
BIOPHARMA

Catalog No. 19167154 Shop All AlphaProTech Products
Change view
Click to view available options
Size:
Universal
X-Large
Cuff Style:
Elastic
Elastic with Thumb Loop
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Cuff Style
19-167-154 Universal Elastic
19-130-4267 X-Large Elastic
19-169-4054 Universal Elastic with Thumb Loop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-167-154 Supplier AlphaProTech Supplier No. MS01J068
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protective sleeves are constructed of a high quality microporous composite material with a low particle count to meet the requirements of controlled environments.

  • Low particle count
  • Tapered sleeve with elastic at both ends
  • Sonic welded seams
  • ComforTech material is made of microporous material for use in cleanrooms and controlled environments

Specifications

Product Type Sleeve Protector
Size Universal
Construction Tapered
Material Microporous Laminate
Color White
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Ultrasonically Welded
Cuff Style Elastic
Latex Free Yes
Model MS-01J06
For Use With (Application) Cleanrooms/Controlled Environments
Certifications/Compliance 21C CFR Part 820
Length (English) 19.5 in.
Length (Metric) 49.5 cm
Packaging Bulk
No. per Case 300
Cleanroom Class Class 10,000 (ISO 7)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.