Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaProTech Critical Cover™ GenPro™ Sleeves
BIOPHARMA

Catalog No. 19165501 Shop All AlphaProTech Products
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-165-501 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-165-501 Supplier AlphaProTech Supplier No. MS011358
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Strong, lightweight, breathable sleeves are designed to protect against contaminants and particulates in non-hazardous environments.

  • For use in non-hazardous environments
  • Elastic on both ends
  • Serged Seams
  • Limited barrier to contaminants and particulates

Specifications

Size Universal
Product Type Sleeve Protector
Construction Tapered
Material Polypropylene
Color White
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Serged
Cuff Style Elastic
Latex Free Yes
Model MS-01135-8
For Use With (Application) Dry, Non-hazardous Environments
Certifications/Compliance 21C CFR Part 820
Length (English) 19.5 in.
Length (Metric) 49.5 cm
Packaging Bulk
No. per Case 200
Cleanroom Class Class 10,000 (ISO 7)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.