Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaProTech Critical Cover™ PFL™ Face Masks Without Magic Arch™
BIOPHARMA

Catalog No. 19160986 Shop All AlphaProTech Products
Change view
Click to view available options
Fastening Type:
Headband
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fastening Type
19-160-986 Headband
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-160-986 Supplier AlphaProTech Supplier No. 9908
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Positive Facial Lock™ PFL face mask features exclusive Twist-Seal™ chin piece to allow a custom fit to any size face, reducing potential for small particles to be inhaled around the borders.

  • Twist-Seal™ chin piece allows a custom fit to any size face
  • latex-free headband
  • Unique PFL design virtually eliminates gaps at the border of the mask
  • Materials used in this 8” PFL mask are fluid resistant and create a durable exterior and a soft interior
  • ASTM F2100 Compliant Level 3 mask

Specifications

Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 3
Color Blue
Fastening Type Headband
Size Standard
Sterility Non-sterile
Filter Efficiency 97.8% (PFE)
Latex Free Yes
Model 9908
Packaging Type Cleanroom Packaged
Pore Size 0.1 μm (PFE)
Resistance Fluid
Series Critical Cover™ PFL™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.