Learn More
AlphaProTech Critical Cover™ PFL™ Face Masks Without Magic Arch™
BIOPHARMA
Catalog No. 191695060 Shop All AlphaProTech Products
Change view
Click to view available options
Fastening Type:
Headband
Catalog No. Fastening Type
19-169-5060 Headband
1 options
This item is not returnable. View return policy
Catalog No. 19-169-5060 Supplier AlphaProTech Supplier No. 108
Add to Cart
Add to Cart
This item is not returnable. View return policy
Positive Facial Lock™ PFL face mask features exclusive Twist-Seal™ chin piece to allow a custom fit to any size face, reducing potential for small particles to be inhaled around the borders.
  • Twist-Seal™ chin piece allows a custom fit to any size face
  • latex-free headband
  • Unique PFL design virtually eliminates gaps at the border of the mask
  • Materials used in this 8” PFL mask are fluid resistant and create a durable exterior and a soft interior
  • ASTM F2100 Compliant Level 3 mask
Specifications
Product Type Medical/Procedural Face Mask
Color Blue
Fastening Type Headband
Sterility Non-sterile
Certifications/Compliance ASTM F2299
Filter Efficiency 97.8% (PFE)
Model 108
Pore Size 0.1 μm (PFE)
Resistance Fluid
Series Critical Cover™ PFL™