Learn More
Sino Biological alpha-Synuclein / SNCA Antibody(HRP), Rabbit PAb, Antigen Affinity Purified 100ug
Supplier:  Sino Biological 12093T16H100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human alpha-Synuclein / SNCA (rh alpha-Synuclein / SNCA; Catalog#12093-HNAE; P37840-1; Met1-Ala140). alpha-Synuclein / SNCA specific IgG was purified by Human alpha-Synuclein / SNCA affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-224-6385