Learn More
ATCC alphaTC1 Clone 9; Pancreatic Adenoma; Mouse (Mus musculus)
Supplier:  ATCC CRL2350
Encompass
alphaTC1 Clone 9 is an alpha cell that was isolated from the pancreas of a mouse with adenoma. This cell line can be used for studying glucagon biosynthesis and alpha cell sensitivity to cytokines.
Catalog No. 50-238-2285