Learn More
MSC Ansell AlphaTec 58-010 Chemical Resistant Gloves, Nitrile, Blue, 1/Pr.
Catalog No. 5000685719
Encompass
These chemical-resistant nitrile gloves are engineered to provide an optimal balance of fit, flexibility, and protection against light acids, greases, oils, and liquids. Designed for all-day comfort, they offer enhanced dexterity and tactility without sacrificing durability.
  • Nitrile construction with 0.28 mm thickness
  • Raised diamond grip design
  • Antistatic and silicone-free composition
  • Gauntlet cuff style for extended forearm protection
  • Latex-free material to prevent Type 1 allergies
  • Compliant with 21 CFR 177.2600 for food contact applications
  • Certified to ANSI/ISEA 105-2024 and various EN standards for safety and protection
Catalog No. 50-006-85719 Supplier MSC Supplier No. 43021286
These chemical-resistant nitrile gloves are engineered to provide an optimal balance of fit, flexibility, and protection against light acids, greases, oils, and liquids. Designed for all-day comfort, they offer enhanced dexterity and tactility without sacrificing durability.
  • Nitrile construction with 0.28 mm thickness
  • Raised diamond grip design
  • Antistatic and silicone-free composition
  • Gauntlet cuff style for extended forearm protection
  • Latex-free material to prevent Type 1 allergies
  • Compliant with 21 CFR 177.2600 for food contact applications
  • Certified to ANSI/ISEA 105-2024 and various EN standards for safety and protection