Learn More
Hanna Pharmaceutical Supply Co., Inc. ALPHATRAK 3 BLOOD GLUCOSE KIT
Supplier:  Hanna Pharmaceutical Supply Co., Inc. 10026571
Encompass_Preferred
monitoring system starter kit, 1 AlphaTRAK® 3 meter, 2 batteries (3 V, CR2032), 50 blood glucose test strips, 1 (4 oz) bottle control solution, 1 lancing device, 30 lancets, 1 diabetes diary, 1 user‘s guide and 1 carrying case
Catalog No. NC2602423