Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alphazurine A, MP Biomedicals™

Catalog No. ICN15473080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15473080 100 g
ICN15473025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15473080 Supplier MP Biomedicals, Inc Supplier No. 0215473080
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Soluble in water (1mg/mL); Store at Room Temperature (15° to 30°C)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3486-30-4
Molecular Formula C37H35N2NaO6S2
Molecular Weight (g/mol) 690.805
MDL Number MFCD00044440
InChI Key FTUYQIPAPWPHNC-UHFFFAOYSA-M
Synonym acid blue 7, alphazurine a, patent blue a, fenazo bluexg, erio glaucine x, hidacid blue a, carmine blue af, hidacid blue af, kiton blue a, patent blue af
PubChem CID 62498
IUPAC Name sodium;4-[[4-[benzyl(ethyl)amino]phenyl]-[4-[benzyl(ethyl)azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzene-1,3-disulfonate
SMILES CCN(CC1=CC=CC=C1)C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC=CC=C4)C=C3)C5=C(C=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+]

Specifications

Color Purple
Quantity 100 g
Formula Weight 690.8
Physical Form Powder
Chemical Name or Material Alphazurine A
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.