Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 249MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: ALPL/597. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Intestine. Cellular Staining: Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human ALPL protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 55kDa.Chromosome: 1p36.12. Gene Symbol: ALPL. Gene ID: 249. Ensemble: ENSG00000162551. Uniprot: NP_016856392.
Catalog No. 50-188-4714