Learn More
ALPAQUA ENGINEERING LLC ALPILLO PLATE CUSHION
Catalog No. NC0050720
Encompass
The Alpillo is an SBS compliant plate support that contains spring cushion technology. The Alpillo gives way as tips push down, thereby compensating for physical tolerances between labware and pipettor that compromises precision pipetting.
Catalog No. NC0050720 Supplier ALPAQUA ENGINEERING LLC Supplier No. A000007
The Alpillo is an SBS compliant plate support that contains spring cushion technology. The Alpillo gives way as tips push down, thereby compensating for physical tolerances between labware and pipettor that compromises precision pipetting.