Learn More
MSC BASF Cockroach Gel Bait, Rotation 2, 4x30 gram, Reservoir
Catalog No. 502837966
Encompass
This cockroach gel bait is part of a rotation system designed to prevent bait aversion in cockroach strains. It contains dinotefuran, an active ingredient with EPA Reduced Risk status, offering fast-acting and reliable control for both gel-bait-averse and non-gel-bait-averse cockroaches. It is packaged as 4x30 gram reservoirs.
  • Helps prevent bait aversion development
  • Contains dinotefuran, an EPA Reduced Risk active ingredient
  • Provides fast kill with excellent secondary nymphal mortality
  • Effective against gel-bait-averse and non-gel-bait-averse cockroaches
  • Offers different food ingredients in a rotation system
  • Kills cockroaches within minutes after exposure
  • Can be rotated with other formulations every six months for optimal aversion management
Catalog No. 50-283-7966 Supplier MSC Supplier No. 74321316
This cockroach gel bait is part of a rotation system designed to prevent bait aversion in cockroach strains. It contains dinotefuran, an active ingredient with EPA Reduced Risk status, offering fast-acting and reliable control for both gel-bait-averse and non-gel-bait-averse cockroaches. It is packaged as 4x30 gram reservoirs.
  • Helps prevent bait aversion development
  • Contains dinotefuran, an EPA Reduced Risk active ingredient
  • Provides fast kill with excellent secondary nymphal mortality
  • Effective against gel-bait-averse and non-gel-bait-averse cockroaches
  • Offers different food ingredients in a rotation system
  • Kills cockroaches within minutes after exposure
  • Can be rotated with other formulations every six months for optimal aversion management