Learn More
MSC BASF Alpine Cockroach Gel Bait, Rotation 2 Reservoir, 4x30 g
Catalog No. 502837965
Encompass
The BASF Alpine Cockroach Gel Bait Rotation 2 Reservoir is formulated for robust cockroach control. This product is engineered as part of a rotation system to combat bait aversion, delivering rapid action and utilizing a reduced-risk active ingredient. Its unique composition effectively attracts both gel-bait-averse and non-gel-bait-averse cockroach strains.
  • Provides fast kill with excellent secondary nymphal mortality.
  • Helps prevent the development of bait aversion in cockroach populations.
  • Appeals to both gel-bait-averse and non-gel-bait-averse cockroaches.
  • Contains dinotefuran, a nonrepellent active ingredient with EPA Reduced Risk Status.
  • Features different food ingredients in each rotation for reliable control.
  • Initiates cockroach elimination within minutes of exposure.
Catalog No. 50-283-7965 Supplier MSC Supplier No. 74320359
The BASF Alpine Cockroach Gel Bait Rotation 2 Reservoir is formulated for robust cockroach control. This product is engineered as part of a rotation system to combat bait aversion, delivering rapid action and utilizing a reduced-risk active ingredient. Its unique composition effectively attracts both gel-bait-averse and non-gel-bait-averse cockroach strains.
  • Provides fast kill with excellent secondary nymphal mortality.
  • Helps prevent the development of bait aversion in cockroach populations.
  • Appeals to both gel-bait-averse and non-gel-bait-averse cockroaches.
  • Contains dinotefuran, a nonrepellent active ingredient with EPA Reduced Risk Status.
  • Features different food ingredients in each rotation for reliable control.
  • Initiates cockroach elimination within minutes of exposure.