Learn More
Gilson Company Inc 200mm Air Jet Test Sieve, All Stainless, No. 60
Catalog No. NC9244029
Encompass_Preferred
200mm diameter Air Jet Test Sieve has stainless steel woven-wire cloth with No. 60 (250µm) opening size and stainless steel frame and is fitted with special sealing gasket.
Catalog No. NC9244029 Supplier Gilson Company Inc Supplier No. AJA228
200mm diameter Air Jet Test Sieve has stainless steel woven-wire cloth with No. 60 (250µm) opening size and stainless steel frame and is fitted with special sealing gasket.