Learn More
TARGETMOL CHEMICALS INC ALPINETIN 25MG
SDP
Catalog No. 502580095
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Alpinetin ((-)-alpinetin) has antibacterial activity.Alpinetin has anti-inflammatory activity.Alpinetin can enhance the sensitivity of HepG2 hepatoma cells to the chemotherapeutic agent CDDP. Alpinetin has strong anti-hepatoma and pancreatic cancer cells effects by inhibiting proliferation regulating of the Bcl-2 family and XIAP expression releasing of cytochrome c and activating caspases. purity: 99%
Catalog No. 50-258-0095 Supplier TARGETMOL CHEMICALS INC Supplier No. T386725MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Alpinetin ((-)-alpinetin) has antibacterial activity.Alpinetin has anti-inflammatory activity.Alpinetin can enhance the sensitivity of HepG2 hepatoma cells to the chemotherapeutic agent CDDP. Alpinetin has strong anti-hepatoma and pancreatic cancer cells effects by inhibiting proliferation regulating of the Bcl-2 family and XIAP expression releasing of cytochrome c and activating caspases. purity: 99%