Learn More
Medchemexpress LLC HY-N0625A 5mg Medchemexpress, Alpinetin CAS:36052-37-6 Purity:>98%
SDP
Catalog No. 501970297
Encompass_Preferred
Medchemexpress, HY-N0625A 5mg Alpinetin CAS:36052-37-6 Alpinetin is a flavonoid isolated from Alpinia katsumadai Hayata, activates activates PPAR-γ, with potent anti-inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0297 Supplier Medchemexpress LLC Supplier No. HYN0625A5MG
Medchemexpress, HY-N0625A 5mg Alpinetin CAS:36052-37-6 Alpinetin is a flavonoid isolated from Alpinia katsumadai Hayata, activates activates PPAR-γ, with potent anti-inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.