Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 250MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM593. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: HepG2 cells. Placenta or seminoma. Cellular Staining: Cytoplasmic, cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human ALPP protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 70kDa.Chromosome: 2q37.1. Gene Symbol: ALPP. Gene ID: 250. Ensemble: ENSG00000163283. Uniprot: NP_001623.
Catalog No. 50-188-4716