Learn More
Aviva Systems Biology Corp ALPP Antibody - C-terminal region (ARP41959_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41959_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ALPP. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ALPP. Alternate names: ALP, PLAP, PALPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58066