Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alprenolol (hydrochloride) | 13707-88-5 | 97.4% | 25 MG
SDP

Catalog No. 5000414004
Encompass_Preferred

Alprenolol hydrochloride is a chemical compound primarily used as a laboratory chemical and for the manufacture of substances. This product is intended for research use only and should be handled by suitably qualified and experienced scientists in appropriately equipped facilities. It is supplied as a white to off-white solid.

  • CAS number: 13707-88-5
  • Molecular formula: C₁₅H₂₄ClNO₂
  • Molecular weight: 285.81
  • Purity: 97.37% (Assay)
  • Appearance: White to off-white solid
  • Identified uses: Laboratory chemicals, manufacture of substances
  • Storage: 4°C, sealed storage, away from light and moisture
  • Hazard warnings: Harmful if swallowed, causes skin irritation, causes serious eye irritation, may cause respiratory irritation

Catalog No. 50-004-14004 Supplier Medchemexpress LLC Supplier No. 25MGHYB1517AR
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alprenolol hydrochloride is a chemical compound primarily used as a laboratory chemical and for the manufacture of substances. This product is intended for research use only and should be handled by suitably qualified and experienced scientists in appropriately equipped facilities. It is supplied as a white to off-white solid.

  • CAS number: 13707-88-5
  • Molecular formula: C₁₅H₂₄ClNO₂
  • Molecular weight: 285.81
  • Purity: 97.37% (Assay)
  • Appearance: White to off-white solid
  • Identified uses: Laboratory chemicals, manufacture of substances
  • Storage: 4°C, sealed storage, away from light and moisture
  • Hazard warnings: Harmful if swallowed, causes skin irritation, causes serious eye irritation, may cause respiratory irritation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.