Learn More
Medchemexpress LLC Alprenolol (hydrochloride) | 13707-88-5 | 99.8% | 1 ML
SDP
Catalog No. 5000411934
Encompass_Preferred
Alprenolol hydrochloride is an orally active non-selective β-adrenoceptor antagonist and an antagonist of 5-HT1A and 5-HT1B receptors. It is used as an anti-hypertensive, anti-anginal, and anti-arrhythmic agent.
  • Orally active non-selective β-adrenoceptor antagonist
  • Antagonist of 5-HT1A and 5-HT1B receptors
  • Functions as an anti-hypertensive agent
  • Functions as an anti-anginal agent
  • Functions as an anti-arrhythmic agent
  • Demonstrates in vivo effects on blood pressure and heart rate in conscious renal hypertensive dogs
  • Effectively blocks anxiolytic effects in adult male Swiss Webster mice
  • Reduces systolic and diastolic pressure and heart rate in cats
Catalog No. 50-004-11934 Supplier Medchemexpress LLC Supplier No. HYB1517A10MM/1ML
Alprenolol hydrochloride is an orally active non-selective β-adrenoceptor antagonist and an antagonist of 5-HT1A and 5-HT1B receptors. It is used as an anti-hypertensive, anti-anginal, and anti-arrhythmic agent.
  • Orally active non-selective β-adrenoceptor antagonist
  • Antagonist of 5-HT1A and 5-HT1B receptors
  • Functions as an anti-hypertensive agent
  • Functions as an anti-anginal agent
  • Functions as an anti-arrhythmic agent
  • Demonstrates in vivo effects on blood pressure and heart rate in conscious renal hypertensive dogs
  • Effectively blocks anxiolytic effects in adult male Swiss Webster mice
  • Reduces systolic and diastolic pressure and heart rate in cats