Learn More
Medchemexpress LLC Alprenolol (hydrochloride) | 13707-88-5 | 99.8% | 100 MG
SDP
Catalog No. 5000411424
Encompass_Preferred
Alprenolol ((RS)-Alprenolol; dl-Alprenolol) hydrochloride is an orally active non-selective β-adrenoceptor antagonist and an antagonist of 5-HT1A and 5-HT1B receptors. It is utilized as an anti-hypertensive, anti-anginal, and anti-arrhythmic agent.
  • Orally active non-selective β-adrenoceptor antagonist.
  • Antagonist of 5-HT1A and 5-HT1B receptors.
  • Used as an anti-hypertensive agent.
  • Used as an anti-anginal agent.
  • Used as an anti-arrhythmic agent.
Catalog No. 50-004-11424 Supplier Medchemexpress LLC Supplier No. HYB1517A100MG
Alprenolol ((RS)-Alprenolol; dl-Alprenolol) hydrochloride is an orally active non-selective β-adrenoceptor antagonist and an antagonist of 5-HT1A and 5-HT1B receptors. It is utilized as an anti-hypertensive, anti-anginal, and anti-arrhythmic agent.
  • Orally active non-selective β-adrenoceptor antagonist.
  • Antagonist of 5-HT1A and 5-HT1B receptors.
  • Used as an anti-hypertensive agent.
  • Used as an anti-anginal agent.
  • Used as an anti-arrhythmic agent.