Learn More
Medchemexpress LLC Alprenolol hydrochloride | 13707-88-5 | 99.8% | 500 MG
SDP
Catalog No. 5000411935
Encompass_Preferred
Alprenolol hydrochloride is an orally active non-selective β-adrenoceptor antagonist and an antagonist of 5-HT1A and 5-HT1B receptors. It is used as an anti-hypertensive, anti-anginal and anti-arrhythmic agent.
  • Orally active non-selective β-adrenoceptor antagonist.
  • Antagonizes 5-HT1A and 5-HT1B receptors.
  • Used as an anti-hypertensive, anti-anginal, and anti-arrhythmic agent.
  • High purity of 99.76%.
  • Soluble in DMSO (100 mg/mL) and water (50 mg/mL).
  • Formulations available for oral and intraperitoneal injection.
Catalog No. 50-004-11935 Supplier Medchemexpress LLC Supplier No. HYB1517A500MG
Alprenolol hydrochloride is an orally active non-selective β-adrenoceptor antagonist and an antagonist of 5-HT1A and 5-HT1B receptors. It is used as an anti-hypertensive, anti-anginal and anti-arrhythmic agent.
  • Orally active non-selective β-adrenoceptor antagonist.
  • Antagonizes 5-HT1A and 5-HT1B receptors.
  • Used as an anti-hypertensive, anti-anginal, and anti-arrhythmic agent.
  • High purity of 99.76%.
  • Soluble in DMSO (100 mg/mL) and water (50 mg/mL).
  • Formulations available for oral and intraperitoneal injection.